Learn More
Troemner™ ASTM Analytical Precision Class 3 Weight Set
Troemner Calibration Weight Sets Adjusted to Meet or Exceed ASTM Class 1 to 4 Tolerances
Supplier: Troemner™ 80859360
Description
- Troemner ASTM Class 1-4 offer high accuracy with tight tolerances and uncertainties of less than 1/4 of the tolerance. Weight surfaces meet or exceed ASTM and have a highly polished finish.
- Available in high grade stainless steels including Troemner Alloy 8 which possesses closely controlled density, extremely low magnetic properties, good stability, and resistance to corrosion.
- Available with certificate options traceable to NIST including ISO/IEC 17025 Accredited Certificate or Traceable Certificate to aid in meeting SOP and regulatory requirements.
Specifications
Calibration Mass | |
Troemner Alloy 8, Stainless Steel | |
Weights 50 kg through 1 g are Cylindrical with Handling Knob; Weights 500 mg and Below are Flat with One End Turned Up For Easy Handling |
No Certificate | |
10 mg to 50 g | |
10 g; (2) 20 g; 100 mg; 10 mg; (2) 2 g; (2) 200 mg; (2) 20 mg; 1 g; 50 g; 5 g; 500 mg; 50 mg |