Learn More
Medicom™ AssureMask Balance™ Procedure Earloop Face Masks, ASTM Level 1
AssureMask Balance™ Procedure Earloop Face Masks, ASTM Level 1 provide healthcare professionals and other frontline workers with the ideal balance between the protection they need and the breathability and comfort they want. Made from high-quality materials.
Supplier: Medicom™ 205415
This item is not returnable.
View return policy
Description
- Shingle pleat design prevents fluid pooling and cross-contamination
- Made from high-quality materials for reliable protection without compromising breathability
- Adjustable nosepiece ensures a comfortable, customized fit
Ultrasonically sealed, soft, stretchy, latex-free earloops prevent pulling and irritation - Available in all 3 levels of ASTM protection
Specifications
Medical/Procedural Face Mask | |
Blue | |
Non-sterile | |
Yes |
ASTM Level 1 | |
Earloops | |
Pleated | |
Yes |