Learn More
Bellco Glass, Inc. The anaerobic culture bottle is designed for the growth of anaerobic bacteria and maintaining anaerobic conditions. The bottle features a 45 gl screw cap with silicone o-ring and blue butyl rubber stopper. This allows for easy access of contents and elimi
SDP
Supplier:  Bellco Glass, Inc. 204850000
Encompass_Preferred
The anaerobic culture bottle is designed for the growth of anaerobic bacteria and maintaining anaerobic conditions. The bottle features a 45 gl screw cap with silicone o-ring and blue butyl rubber stopper. This allows for easy access of contents and elimi
Catalog No. 50-194-5349