Learn More
Sigma Aldrich Fine Chemicals Biosciences Aspirator tube assemblies for calibrated microcapillary pipettes, 5/pack
Supplier: Sigma Aldrich Fine Chemicals Biosciences A51775EA
Consists of flexible silicone rubber nosepiece, hard plastic mouthpiece, and 15 inches of latex tubing.