Learn More
Newcomer Supply Fungus, GMS, Aspergillus sp., Artificial Control Slides, 98/set
Supplier: Newcomer Supply 4220B
Newcomer Supply Fungus, GMS, Aspergillus sp., Artificial Control Slides are for the positive histochemical staining of Aspergillus sp. in tissue sections. Aspergillus fumigatus purchased from Remel Microbiology Products is used to produce the positive control tissue. Fungus, GMS, Aspergillus sp., Artificial Control Slides contain sections of positive staining rat lung and negative staining human lung.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.