Learn More
Kamiya Biomedical ASO/RF/CRP Control, L1: 2 x 2mL, L2: 2 x 2mL RX
Supplier: Kamiya Biomedical K55C4M
K-ASSAY ASO/RF/CRP Control. Level 1: 2 x 2 mL (lyophilized), Level 2: 2 x 2 mL (lyophilized). The K-ASSAY ASO/RF/CRP Control is intended for use as an assayed quality control material for monitoring the performance of the K-ASSAY Anti-Streptolysin O (ASO, Cat. No. KAI-078), Rheumatoid Factor (RF (Ver.2), Cat. No. KAI-230), C-Reactive Protein (CRP, Cat. No. KAI-026), and wide range CRP (Cat. No. KAI-082) immunoturbidimetric assays. FOR IN VITRO DIAGNOSTIC USE.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.