Learn More
Kamiya Biomedical ASO Calibrator, 5x1mL/PK
Supplier: Kamiya Biomedical KAI079C
K-ASSAY ASO (WHO) Calibrator. 5 x 1 mL (liquid). For 5 point calibration. The ASO Calibrators have been standardized against the WHO (World Health Organization) NIBSC Anti-Streptolysin O standard material. The K-ASSAY ASO (WHO) Calibrator is intended to be used for the calibration of the K-ASSAY ASO (WHO) immunoturbidimetric assay (Cat. No. KAI-078). FOR IN VITRO DIAGNOSTIC USE.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.