Learn More
Bioassay Systems EnzyChrom™ Ascorbic Acid Assay Kit. For quantitative determination of ascorbate (ascorbic acid) and evaluation of drug effects on ascorbic acid metabolism.
Supplier: Bioassay Systems EASC100
For quantitative determination of ascorbate (ascorbic acid) and evaluation of drug effects on ascorbic acid metabolism. Key Features: Use 20 µL samples. Linear detection range: colorimetric assay 6 to 1000 µM, fluorimetric assay 1 to 100 µM ascorbic acid. Method: OD570nm, or FL530/585nm. Samples: Serum, plasma, urine, saliva, milk, tissue, and cell culture. Species: All. Procedure: Assay takes 10 min. Kit size: 100 tests. Detection limit: OD, FL: 6, 1 µM.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.