Learn More
MilliporeSigma™ Supelco™ Ascentis™ Express OH5 UHPLC Column, 2 μm Particle Size
The Ascentis™ Express 2 μm OH5 stationary phase is a highly polar ligand that possesses 5 hydroxyl groups tethered to the silica via novel proprietary linkage chemistry.
Supplier:  MilliporeSigma™ Supelco™ 50958U
 View more versions of this product
Catalog No. 11-100-7079
Add to Cart
Description
Description
The Ascentis™ Express 2 μm OH5 stationary phase is a high performance material that provides a UHPLC column that can be used for traditional normal phase separations using non-polar, totally organic mobile phases or for aqueous normal phase chromatography with the typical mobile phases for hydrophilic interactive liquid chromatography (HILIC) of basic, acidic, zwitterionic, or neutral compounds.
Specifications
Specifications
2.1 mm
2 μm
Hydroxy, Penta-
Hydroxy, Penta- Phase/Fused-Core Particle Platform
Reversed Phase
Fused-Core UHPLC
7.5 cm
90 Å
Ascentis
Yes
LC-MS Column
Product Suggestions
Product Suggestions
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Ascentis is a registered trademark of Sigma-Aldrich Co. LLC