Learn More
MilliporeSigma™ Supelco™ Ascentis™ Express HILIC UHPLC Column, 2 μm Particle Size
Contains a bare silica stationary phase that can be used for traditional normal phase separations or aqueous normal phase chromatography.
Supplier:  MilliporeSigma™ Supelco™ 51409U
 View more versions of this product
Catalog No. 11-100-7002
Add to Cart
Description
Description
The Ascentis™ Express 2 μm HILIC or bare silica stationary phase provides a UHPLC column that can be used for traditional normal phase separations using non-polar, totally organic mobile phases or for aqueous normal phase chromatography with the typical reversed-phase mobile phases for hydrophilic interactive liquid chromatography (HILIC) of basic, acidic, or neutral compounds.
Specifications
Specifications
2.1 mm
2 μm
Silica
Silica Phase/Fused-Core Particle Platform
Reversed Phase
10 cm
90 Å
Ascentis
No
Fused-Core UHPLC
Product Suggestions
Product Suggestions
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Ascentis is a registered trademark of Sigma-Aldrich Co. LLC