Learn More
Showa Denko America Inc ASAHIPAK NH2P-50 4E 250X4.6 5
Supplier: Showa Denko America Inc F7630001
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Polyvinyl alcohol based amino HILIC series (hydrophilic Interaction Chromatography). Durable with no shedding, these columns are excellent for light scattering and charged aerosol detectors.