Learn More
Bioworld ARTOCARPUSJACALIN) - LECTIN
Supplier: Bioworld 215100011
Artocarpus integrifolia (Jacalin) Lectin isolated from Artocarpus integrifolia (jackfruit) seeds. Specifications: Primary sugar specificity: Galactose Preferential binding activity: galactosyl (beta-1,4) N-acetylglucosamine.