Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ ART4 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Supplier: Abnova™ H00000420T01
Specifications
Western Blot | |
ART4 293T Cell Transient Overexpression Lysate(Denatured) | |
Theoretical M.W. 34.65kDa | |
Transient overexpression cell lysate was tested with anti-ART4 antibody (Mfr. No. H00000420-B01) by Western Blot. | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
293T | |
NM_021071.2 | |
pCMV-ART4 full-length | |
100 μL | |
Human |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction