Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ergodyne™ Arsenal™ 5116 General Duty Gear Bag
Arsenal™ 5116 General Duty Gear Bag is designed to store, organize, and easily carry everything you need for the workday or weekend.
$110.10
Specifications
Material | 600D Polyester |
---|---|
Dimensions (L x W x H) | 23.5 x 9.5 x 12 in. (60 x 24 x 30 cm) |
For Use With (Application) | Contractors, Construction, Electricians, General Purpose |
Quantity | 1 Ea. |
Size | Medium |
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
19-819-720
|
Ergodyne™
13016 |
Black |
Each for $110.10
|
|
|||||
19-819-823
|
Ergodyne™
13416 |
Realtree Xtra™ | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Durable material withstands wear and tear
- 1680D ballistic polyester base reinforces the bottom for ultimate durability
- Water-resistant backing helps to repel water and stains
- Heavy-duty webbing straps on top and side of duffel for extra reinforcement
- Removable and adjustable padded shoulder strap for easy transportation
- Heavy-duty zippers to secure gear inside the duffel
- Nickel-plated Storm Drains™ added to each compartment for ventilation
- Two large side pockets for cell phones, headphones, pens, keys, and other small parts
Specifications
600D Polyester | |
Contractors, Construction, Electricians, General Purpose | |
Medium |
23.5 x 9.5 x 12 in. (60 x 24 x 30 cm) | |
1 Ea. | |
44 L |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title