Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Arlington Scientific Storage Compartment for ASI Hampton Lounge

Accessory for Arlington Scientific Hampton Lounge
Supplier: Arlington Scientific 20617010
Description
For use with Arlington Scientific Hampton Lounge
Specifications
Hampton lounges | |
Storage Compartment |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title