Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sartorius Arium™ Smart Station UP
Arium™ Smart Station flexible remote dispenser for ultrapure water
$3316.00
Specifications
Capacity (Metric) | 50 mL to 50 L |
---|---|
Feedwater Source | Ultrapure |
Product Water Type | Ultrapure (Type 1) |
Display | Touch display |
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
14-556-150
|
Sartorius
H2OARSTUPT |
Smart Station Benchtop |
Each for $3,316.00
|
|
|||||
14-556-151
|
Sartorius
H2OARSTUPB |
Smart Station Wall-Mounted |
Each for $3,316.00
|
|
|||||
Description
- Height adjustable display/dispense station for dispensing Type 1 ultrapure water
- Compact design for easy and space-saving integration into lab environment
- Intuitive touch-activated color display with direct access to important functions
- Flexible and stepless height adjustment to fill different sized containers and beakers
- Ergonomic design can be optimized for right or left hand operation
- Accurate volume dispense for fast and precise buffer and sample prep
- Adjustable, splash-free flow rate from drop-by-drop to 2 L/min
Equipment supplied:
- Display/dispense unit with stand
- Power supply including:
- IEC connector
- Connection set
For intended use with:
- Arium™ Bagtank
- Arium™ Pro
- Arium™ Comfort
Specifications
50 mL to 50 L | |
Ultrapure | |
Ultrapure (Type 1) | |
Touch display |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title