Learn More
Cole-Parmer Argos Technologies Pipette Basins, 12 Channel x 3 mL, Sterile, Individually Wrapped; 20/Cs
Supplier: Cole-Parmer 0439533
The pipette reagent reservoirs feature a V-shaped bottom, making multichannel pipetting easier and ensuring complete aspiration of costly reagents. The 12-Chamber Reservoir is designed for serial dilutions or pipetting different reagents using a 12-channel pipettor. Each numbered chamber holds up to 3 mL. The chemical-resistant polypropylene reservoir can withstand temperatures from -122 to 250F (-80 to 121C). Tight fitting lid minimizes evaporation or contamination during incubation or storage.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.