Learn More
Epredia™ ArcosSL™ Trays and Storage
Enhance traceability and organization with trays, storage cabinets and cardboard sleeves for the Epredia™ ArcosSL™ slide management system.
Supplier: Epredia™ A85310011
This item is not returnable.
View return policy
Description
Smart and Secure Storage- Each Epredia™ ArcosSL™ tray can hold up to 240 slides and features a unique serial number and easy-to-navigate XY coordinate guides. Slides are neatly arranged in the tray, rapidly scanned, and archived in sturdy cabinets, making them easy to identify and locate. Each cabinet can hold up to 40 trays and can be easily configured to fit your facility needs.
- ArcosSL trays hold slides securely in a shingled manner, allowing maximum visibility to each slide’s tab while maintaining a separation between the slides to avoid damage and cross contamination. This allows you to optimize tray space and eliminate gaps.
- ArcosSL cabinets offer flexible storage and easy retrieval of slides in a secure fashion. They can be stacked two units high and safety requirements such as the cabinet base (single or double), bar and fixation ensure your slide’s stability during storage.
- Cardboard sleeves may be used as an alternative storage option in place of the ArcosSL cabinets. Each sleeve can hold up to four ArcosSL trays.
Specifications
ArcosSL™ cabinet, includes 40 trays |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Epredia™ ArcosSL™ instrumentation requires the use of ArcosSL trays, sold separately. ArcosSL cabinets are highly recommended for storage.
Single cabinet base is required for single ArcosSL cabinet configuration.
Double cabinet base is required for double ArcosSL cabinet configuration.
ArcosSL cabinet fixation is a safety requirement for ArcosSL cabinets.
ArcosSL cabinet bar is a safety requirement for ArcosSL cabinets.