Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AQUASTAR™ Anhydrous Methanol, MilliporeSigma™

Click to view available options
Amber Glass; 4L
This item is not returnable. View return policy
This item is not returnable. View return policy
  • Methanol for Volumetric Karl Fischer Titrations.
  • For moisture determination. Assay >99.8%. Max. 0.01% H&subscript_2;O content.
Color Colorless to Yellow
Quantity 4 L
CAS 67-56-1
Packaging Amber Glass
Physical Form Liquid
Molecular Formula CH4O
Linear Formula CH3OH
MDL Number MFCD00004595
UN Number 1230
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name methanol
Molecular Weight (g/mol) 32.04
Formula Weight 32.04
Chemical Name or Material AQUASTAR™ Anhydrous Methanol, AQUASTAR™
Show More Show Less

DOTInformation : DOT Class 3, : Flammable Liquid, Toxic

Recommended Storage : Room Temperature

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit