Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OHAUS™ Aquasearcher™ Multi-Parameter Analyzer AB33M1

The Aquasearcher™ Multi-Parameter Analyzer AB33M1 is easy-to-use and accurate, capable of measuring up to seven parameters including pH, ORP, conductivity, TDS, salinity, resistivity, and temperature.
$925.00 - $1080.00
Specifications
Channels | pH, Conductivity |
---|---|
Display Type | LCD |
Measurement Modes | Auto-stop, Continuous |
Outputs | RS232, USB Device (Included) |
Range (Conductivity) | 0.01 μS/cm to 500 mS/cm |
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
S05299
![]() |
OHAUS™
30589824/EMD |
AB33M1 Meter, Electrode Holder |
Each for $925.00
|
|
S05298
![]() |
OHAUS™
30589825/EMD |
AB33M1 Meter, ST310 Electrode, STCON3 Probe, Electrode Holder, 4 pH Buffer Mini Kits (50 mL), Standard Conductivity Solution Kits (1413 μS/cm, 50 mL) |
Each for $1,080.00
|
|
Description
- Features two independent measuring channels which can work simultaneously
- Equipped with a large 6.5 in. LCD display and a touch keypad
- Features a 1000 item memory for measurements and calibration trail
- Provides connectivity and data output capabilities with RS232 and USB interface
- i-Steward monitors the condition of electrodes to ensure repeatable and consistent measurement accuracy
- Intuitive user interface makes changing the parameters, executing setup, and performing calibration as simple as using a smartphone
- Choose between just the AB33M1 meter with electrode holder and a full kit including AB33M1 meter, ST310 electrode, STCON3 probe, electrode holder, 4 pH buffer mini kits (50 mL), standard conductivity solution kits (1413 μS/cm, 50 mL)
Specifications
pH, Conductivity | |
Auto-stop, Continuous | |
0.01 μS/cm to 500 mS/cm | |
-2 to 20 | |
0 to 100 psu | |
0.1/0.01 pH |
LCD | |
RS232, USB Device (Included) | |
-2000 to 2000 mV | |
2 to 100 MΩ-cm | |
0.1 mg/L to 199.9 g/L | |
-5°C to +110°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title