Learn More
Cole-Parmer Stuart Aquatron Water Still, 4 LPH, 230 VAC
Supplier:  Cole-Parmer 9929405
Encompass_Preferred
The Aquatron cabinet-style water stills are available in single or double distilled water models. The double distilled model features higher purity levels. The stills are made of borosilicate glassware coupled with silica-sheathed heaters and then enclosed in a protective cabinet, delivering pyrogen-free distillate of the highest purity from virtually any raw water supply. Its condenser is designed to make sure droplets of distilled water remain in contact with the cooling coil for an extended period, ensuring maximum energy transfer and production of cold distilled water for immediate use. The all-glass construction allows for rapid descaling using strong mineral acids. The units can be safely and easily cleaned without dismantling the glassware. Also, a large bore stopcock with PTFE key is fitted for easy draining of chemicals, after cleaning. Its safety features include a flow sensor, overtemperature thermostat and acrylic screen, along with a cabinet lid and more.
Catalog No. 50-196-0450