Learn More
Cole-Parmer Applikon Fermentation Septum Needle; Stainless Steel
Supplier:  Cole-Parmer 0191920
Encompass_Preferred
Septum needle is for dosing liquids or adding chemicals into your fermentation culture. Needle has a 3/16" OD stainless steel pipe for optional tubing hookup. Autoclavable.
Catalog No. NC1949662
Due to product restrictions, please to purchase or view availability for this product.