Learn More
Electron Microscopy Sciences Willco-dish Glass Bottom Kit Assembly Device 60x20 mm, 40 mm Glass Aperture
Supplier: Electron Microscopy Sciences 7066522
Round, anodized, aluminum “Assembly Devices" for a quick, accurate and safe positioning and assembly of the glass coverslip and the adhesive ring. 60x20 mm.
Aperture: 40 mm