Learn More
Biolegend APC anti-mouse CD38 [S21016F]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Supplier: Biolegend 165604
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
APC anti-mouse CD38 [S21016F]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug