Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zhangjiagang Huachang Pharmaceutical Co., Ltd. AOF L-Lysine
L-Lysine
Supplier: Zhangjiagang Huachang Pharmaceutical Co., Ltd. 99141823
Description
- Appearance: White or Pail Yellow Powder
- Assay: 98.5 to 102.0%
- Solubility: 10% solution is clear, complete and colorless to yellow.
- Loss of drying: ≤5.0%
- Residue on Ignition: ≤0.50%
- Total microbial count: ≤50 CFU/Gram
- Endotoxin: <20 EU/g
- Animal Origin Free
Specifications
25 kg | |
<20 EU/g | |
Powder | |
White or Pale Yellow |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction