Learn More
TOKU-E Streptomycin ReadyMade™ Solution, 3810-74-0, 10x1ML
Supplier: TOKU-E S14210X1ML
Antibiotic, ReadyMade™, sterile-filtered, 1 mg/ml in 1mM EDTA. Useful in cancer research, microbiology, plant biology.