Learn More
Grainger Vibration Isolation Pad, Neoprene, 800 lb Max. Steady Load, 4 in Length, 4 in Width
Supplier: Grainger 4C977
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Pads protect equipment from shaking and are designed to be used when machines are bolted to the floor.