Learn More
NOTRAX™ Anti-Stat P.O.P.™ Vinyl Work Surface
Anti-Stat P.O.P.™ is a vinyl work surface that dissipates static from conductive items placed on it
$794.00 - $922.50
Catalog Number | Mfr. No. | Length (English) | Width (English) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (English) | Width (English) | Price | Quantity & Availability | ||||
19-086-187
|
NOTRAX™
827R3040BU |
30 in. | 40 ft. |
Each for $794.00
|
|
||||
19-086-188
|
NOTRAX™
827R3640BU |
3 ft. | 40 ft. |
Each for $922.50
|
|
||||
19-086-184
|
NOTRAX™
827C0024BU |
Up to 60 ft. | 2 ft. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
19-086-185
|
NOTRAX™
827C0030BU |
Up to 60 ft. | 30 in. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
19-086-186
|
NOTRAX™
827C0036BU |
Up to 60 ft. | 3 ft. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
19-086-189
|
NOTRAX™
827R3660BU |
3 ft. | 60 ft. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
- Protects table and keeps sensitive components safe from damaging static shocks
- Smooth top surface is abrasion resistant and easy to wipe clean
- ESD Static Dissipative
- Meets EOS/ESD-S4, measured resistance for work surfaces RG 107 Ω 10 and RP 108 Ω
- Electrostatic charge (walking test), meets ISO6356 and EN1815
- Grounding cord available, not required
Specifications
Blue | |
1 Ea. | |
1/4 in. |