Learn More
Advanced ImmunoChemical, Inc. Monoclonal Mouse Anti-Human Proinsulin, 0.2mg
Supplier: Advanced ImmunoChemical, Inc. 1PINS0.2MG
This item is manufactured by HyTest, Ltd and distributed by Advanced ImmunoChemical, Inc.
Monoclonal antibodies / Metabolic Syndrome / Monoclonal mouse anti-human proinsulin. MAbs: 3A1, 1G4, 1D6; Hybridoma clones have been derived from hybridization of X63-Ag8-653 myeloma cells with spleen cells of Balb/c mice immunized with purified human proinsulin
Specificity: MAbs 3A1 and 1G4 are directed to different antigenic determinants. MAb 3A1 reacts with the site of connection of C-peptide and C-end of B-chain of proinsulin molecule. MAbs 1D6 reacts with different epitopes on human proinsulin molecule; both of each are generated by the interactions between insulin and C-peptide parts of prohormone. Kd for MAb 3A1 is 1.1 x 10-8 M.Kd for MAb 1G4 is 2.6 x 10-7 M. Kd for MAb 1D6 is >3 x 10-8 M. MAbs 3A1, 1G4, 1D6 do not cross-react with human, bovine and porcine insulin, bovine and porcine proinsulin, and human C-peptide.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.