Learn More
NEDD4 binding protein 2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant N4BP2.
Supplier: Abnova Corporation H00055728A01
Description
This gene encodes a protein containing a polynucleotide kinase domain (PNK) near the N-terminal region, and a Small MutS Related (Smr) domain near the C-terminal region. The encoded protein can bind to both B-cell leukemia/lymphoma 3 (BCL-3) and neural precursor cell expressed, developmentally downregulated 4, (Nedd4) proteins. This protein binds and hydrolyzes ATP, may function as a 5'-polynucleotide kinase, and has the capacity to be a ubiquitylation substrate. This protein may play a role in transcription-coupled DNA repair or genetic recombination. [provided by RefSeq]
Sequence: HLAAIEIFEKVNASLLPQNVLDLHGLHVDEALEHLMRVLEKKTEEFKQNGGKPYLSVITGRGNHSQGGVARIKPAVIKYLISHSFRFSEIKPGCLKVMLKSpecifications
NEDD4 binding protein 2 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant N4BP2. | |
N4BP2 | |
B3BP/FLJ10680/KIAA1413 | |
Mouse | |
50 μL | |
Yes | |
55728 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_018177 | |
N4BP2 | |
N4BP2 (NP_060647, 1671 a.a. ∼ 1770 a.a) partial recombinant protein with GST tag. | |
RUO | |
Primary | |
Human | |
Serum |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.