Learn More
PBL Assay Science Anti-Mouse Interferon Alpha (IFN-Alpha) Rabbit Polyclonal Antibody (PAb) [2e4 (20,000) units/vial]
Supplier: PBL Assay Science 321001
Rabbit polyclonal antibody Anti-Mouse Interferon Alpha is manufactured by PBL Assay Science (Catalog #32100-1) for research use only (RUO) and packaged in 2e4 (20,000) units/vial. This PAb binds to and neutralizes mouse IFN alpha. It is suitable for use in neutralization, immunohistochemistry, immunoprecipitation, and ELISA applications.