Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
prostaglandin reductase 1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant LTB4DH.
Supplier: Abnova Corporation H00022949A01
Description
This gene encodes an enzyme that is involved in the inactivation of the chemotactic factor, leukotriene B4. The encoded protein specifically catalyzes the NADP+ dependent conversion of leukotriene B4 to 12-oxo-leukotriene B4. A pseudogene of this gene is found on chromosome 1. Alternative splicing results in multiple transcript variants. [provided by RefSeq]
Sequence: MKKFGRIAICGAISTYNRTGPLPPGPPPEIVIYQELRMEAFVVYRWQGDARQKALKDLLKWVLEGKIQYKEYIIEGFENMPAAFMGMLKGDNLGKTIVKSpecifications
prostaglandin reductase 1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant LTB4DH. | |
PTGR1 | |
LTB4DH/MGC34943/ZADH3 | |
Mouse | |
50 μL | |
Yes | |
22949 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_012212 | |
PTGR1 | |
LTB4DH (NP_036344, 230 a.a. ∼ 328 a.a) partial recombinant protein with GST tag. | |
RUO | |
Primary | |
Human | |
Serum |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction