Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin Β1 Mouse anti-Human, Clone: 6S6, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB2253
Description
Detect Integrin β1 using this Anti-Integrin β1 Antibody, clone 6S6 validated for use in flow cytometry, immunohistochemistry and immunoprecipitation.Specifications
Integrin β1 | |
Monoclonal | |
Unconjugated | |
NM_002292.3 | |
Mouse | |
Protein A purfied | |
Primary | |
2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
IgG1 |
Flow Cytometry, Immunohistochemistry, Immunoprecipitation | |
6S6 | |
Protein A Purified mouse immunoglobulin in 20mM sodium phosphate, 250mM NaCl, pH. 7.6, with 0.1% sodium azide as a preservative. | |
LAMB2, S-laminin, LAMS | |
Human synovial cells | |
100 μg | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction