Learn More
Advanced ImmunoChemical, Inc. Monoclonal Mouse Anti-Rat-Mouse Insulin/Proinsulin, 0.2mg
Supplier: Advanced ImmunoChemical, Inc. 1IP0.2MG
This item is manufactured by HyTest, Ltd and distributed by Advanced ImmunoChemical, Inc. Monoclonal antibodies / Metabolic Syndrome / Monoclonal mouse anti-rat-mouse insulin/proinsulin. MAbs:D6C4, D3E7Specificity:MAbs D6C4 and D3E7 directed to different antigenic determinants. Kd for MAb D6C4 is 8.1 x 10-8 M.Kd for MAb D3E7 is 6.3 x 10-8 M.MAb D6C4 and D3E7 cross-react with human, bovine and porcine insulin and proinsulin.MAb Isotypes:IgG1, kappa for MAbs D6C4 and D3E7Applications:Detection of rat-mouse insulin/proinsulin in two-site enzyme immunoassay. Recommend pair (capture-detection): D6C4 – D3E7.Antibodies D6C4 and D3E7 can be used in immunohistochemistry on frozen sections.Purification:Chromatography on protein A SepharosePresentation:PBS, pH 7.4, 0.1 % sodium azide (NaN3)Storage Conditions:+ 4 °CMaterial Safety Note:This product is sold as an antibody preparation for research use only. Standard Laboratory Practices should be followed when handling this material.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.