Learn More
Impacto™ Anti-Impact Glove with Wrist Support
Impacto™ 704-20 Glove with padding in the palm and thumb web area protects from impact and vibration and wrist support protects from RSI
Supplier: Impacto™ 70420120020
Description
Impacto™ Anti-Impact Glove with Wrist Support
- Fingerless nylon Lycra and grain leather glove
- VEP padding in the palm and thumb web area
- Detachable grain leather wrist support minimizes extension and over flexion of the wrist to prevent RSI such as carpal tunnel syndrome or tendinitis
- Provides protection from abrasion, impact and vibration
- Fingerless style for optimum dexterity
- Used with hand or electric tools in repetitive tasks
Certifications
EN 420:2003 + A1:2009; EN 388:2003 2243
Specifications
Anti-impact gloves | |
22.86 cm | |
Small/7 | |
Nylon, Lycra |
Fingerless | |
9 in. | |
Hand and electric tools with repetitive tasks |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.