Learn More
anti-IgG2a κ, Mouse, Clone: G155-178, NA/LE, Isotype Control, BD
Supplier: BD Biosciences 554645
Description
The G155-178 clone has an unknown specificity. Trinitrophenal (TNP), the immunogen, is a hapten not expressed on human, mouse, rat or non-human primate cells. In the absence of specific binding, this antibody may bind non-specifically to Fc receptors. The immunoglobulin from clone G155-178 was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by functional assay and isotype control. Other applications were tested at BD Biosciences Pharmingen during
antibody development only or reported in the literature.
Host Species: Mouse
Clone: G155-178
Isotype: IgG2a, κ
Immunogen: TNP-keyhole limpet hemocyanin
ELISA
Specifications
ELISA | |
1mg/mL | |
Mouse | |
Affinity Purified | |
Undiluted at 4°C |
G155-178 | |
Unconjugated | |
TNP-keyhole limpet hemocyanin | |
0.5 mg | |
IgG2a κ |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.