Learn More
anti-IgG1 κ, Mouse , PE-Cy™5, Clone: MOPC-21, BD
Supplier: BD Biosciences 557224
Description
The MOPC-21 immunoglobulin is a mouse myeloma protein. The MOPC-21 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This reagent has been used as isotype control in immunophenotypic studies with baboon, rhesus, or cynomolgus peripheral blood cells. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: MOPC-21
Isotype: IgG1 κ
Species Reactivity: Baboon/Rhesus/Cynomolgus
Flow Cytometry, Isotype Control
Specifications
Flow Cytometry | |
PE-Cyanine5 | |
Affinity Purified | |
Baboon, Cynomolgus, Rhesus Monkey | |
IgG1 κ |
MOPC-21 | |
Mouse | |
50 Tests | |
Undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.