Learn More
anti-IgG1 κ, Mouse, APC, Clone: MOPC-21, Isotype Control, BD
$158.00 - $190.00
Specifications
Clone | MOPC-21 |
---|---|
Concentration | 0.2mg/mL |
Applications | Flow Cytometry, Western Blot |
Conjugate | APC |
Host Species | Mouse |
Description
The MOPC-21 immunoglobulin is a mouse myeloma protein. The MOPC-21 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: MOPC-21
Isotype: IgG1 κ
Species Reactivity: Non-human primate
Flow Cytometry, Intracellular Staining, Isotype Control
Specifications
MOPC-21 | |
Flow Cytometry, Western Blot | |
Mouse | |
IgG1 κ | |
Undiluted at 4°C |
0.2mg/mL | |
APC | |
Non-human Primate | |
Affinity Purified |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.