Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-IgG1 κ Mouse, APC, Clone: MOPC-21, Isotype Control, BD
Supplier: BD Biosciences 555751
Description
The MOPC-21 immunoglobulin is a mouse myeloma protein. The MOPC-21 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: MOPC-21
Isotype: IgG1, κ
Flow Cytometry, Isotype Control
Specifications
Flow Cytometry | |
APC | |
Affinity Purified | |
Undiluted at 4°C. Protect from exposure to light. Do not freeze. |
MOPC-21 | |
Mouse | |
100 Tests | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction