Learn More
Advanced ImmunoChemical, Inc. Monoclonal Mouse Anti-Infectious Bronchitis Virus (IBV), Mabs:IB95, 0.2mg
Supplier: Advanced ImmunoChemical, Inc. 5IBV0.2MG
This item is manufactured by HyTest, Ltd and distributed by Advanced ImmunoChemical, Inc. Monoclonal Antibodies / Veterinary / Monoclonal mouse anti-infectious bronchitis virus (IBV). MAbs:IB95Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells ofBalb/c mice immunized with semi-purified inactivated IB virus of Massachusetts type from infected chicken embryos.Specificity:Nucleoprotein of IBV.MAb Isotypes:IgG2a for MAb IB95Applications:Immunodetection of IBV N-protein in Western Blotting; IB virus detection in sandwich-type immunoassay (capture).Purification:Chromatography on protein A SepharosePresentation:PBS, pH 7.4, 0.1 % sodium azide (NaN3)Storage Conditions:+ 4 °CMaterial Safety Note:This product is sold as an antibody preparation for research use only. Standard Laboratory Practices should be followed when handling this material.Contains sodium azide (0.1 %) as preservative.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.