Learn More
PBL Assay Science Anti-Human Interferon Beta (IFN-Beta) Rabbit Polyclonal Antibody (PAb) [2e4 (20,000) units/vial]
Supplier: PBL Assay Science 314101
Rabbit polyclonal antibody Anti-Human Interferon Beta is manufactured by PBL Assay Science (Catalog #31410-1) for research use only (RUO) and packaged in 2e4 (20,000) units/vial. This PAb binds to and neutralizes human IFN beta. It is suitable for use in neutralization, immunohistochemistry, immunoprecipitation, ELISA, and Western Blot applications.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.