Learn More
Vector Laboratories Horse Anti-Goat, DyLight 488 DI-3094-1.5
SDP
Supplier:  Vector Laboratories DI30941.5
Encompass_Preferred
Horse Anti-Goat IgG Antibody (H+L), DyLight 488, 1.5 mg, is an affinity-purified, ultrapure, high-affinity antibody that has been thoroughly adsorbed against serum and immunoglobulins from potentially interfering species. Unless otherwise specified, antibodies recognize both heavy and light chains (H+L). Optimally labeled with DyLight 488 to provide the brightest label for fluorescence microscopy. Supplied in solution. Excitation: 493 nm; Emission: 518 nm; Color: Green.
Catalog No. 50-204-5641