Learn More
Vector Laboratories Goat Anti-Human Lambda, FL BP-9500-50
SDP
Supplier:  Vector Laboratories BP950050
Encompass_Preferred
Goat Anti-Human Lambda Chain Antibody, Fluorescein, 0.5 mg, is prepared by hyperimmunizing animals in a manner that produces high-affinity antibodies. These are then purified by affinity chromatography to remove any low-affinity antibodies and by solid-phase adsorption techniques to remove cross-reactivities. The final product is subjected to rigorous QC, including immunodiffusion, solid-phase enzyme immunoassays, gel electrophoresis, and solid-phase binding assays. Conjugation ensures the maximum degree of labeling without compromising antibody specificity or affinity. The labeled antibody then undergoes further QC, including immunohistochemical analysis. This chain-specific antibody is produced specifically to distinguish between chains or classes of target immunoglobulins and has virtually no cross-reactivity with other immunoglobulin classes or other heavy or light chains. Fluorescein has an excitation maximum of 495 nm and an emission maximum of 515 nm.
Catalog No. 50-204-5625