Learn More
CD45RO Mouse anti-Human, APC, Clone: UCHL1, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 559865
Description
CD45RO, a member of the CD45 family that includes CD45, CD45RA, and CD45RB, recognizes a 180-kilodalton (kd) isoform of the leucocyte common antigen (LCA). The CD45 antigen is a protein tyrosine phosphatase. In peripheral blood, the CD45RO antigen is present on approximately 40% of resting peripheral blood T lymphocytes, including the CD4+ and CD8+ subpopulations, as well as on most thymocytes and activated T lymphocytes. It is also expressed on monocytes, macrophages, and granulocytes. The CD45RO antigen is present at low density early in the T-lymphocyte maturation cycle. Upon activation by phytohemagglutinin (PHA) or alloantigen, naive T lymphocytes first acquire CD45RO and then lose CD45RA. When these activated T lymphocytes are rechallenged, the cells that exhibit a secondary response are primarily CD45RO+ , leading to the concept that CD45RO+ cells are a primed population of memory T lymphocytes.
Specifications
CD45RO | |
Monoclonal | |
APC | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
UCHL1 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human IL-2-dependent T-cell line | |
100 Tests | |
Immunology | |
Human | |
IgG2a κ |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.