Learn More
CD3ε Mouse anti-Pig, FITC, Clone: BB23-8E6-8C8, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 559582
Description
The BB23-8E6-8C8 monoclonal antibody specifically binds to the 25-kDa ε chain of the T-cell receptor-associated CD3 complex. It recognizes all CD4+ and most CD8+ peripheral blood T lymphocytes, most thymocytes and phytohemagglutinin-stimulated blasts, and subsets of spleen and Peyer’s patch lymphocytes. BB23-8E6-8C8 is a immunoglobulin isotype switch variant of the BB23-8E6 clone. This isotype-switch variant induces a proliferative response of peripheral blood mononuclear cells. The epitope recognized by BB23-8E6 mAb was designated CD3a by the Second International Swine CD Workshop.
Specifications
CD3 ε | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Pig peripheral blood mononuclear cells | |
0.1 mg | |
Immunology | |
Porcine | |
IgG2a κ |
Flow Cytometry | |
BB23-8E6-8C8 | |
FITC | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.