Learn More
CD107a Mouse anti-Human, APC, Clone: H4A3, BD
Mouse Monoclonal Antibody
$222.00 - $632.00
Specifications
Antigen | CD107a |
---|---|
Clone | H4A3 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | APC |
Description
Reacts with the heavily glycosylated 110kDa lysosomal-associated membrane protein, LAMP-1. LAMP-1 is a widely expressed intracellular antigen. It is also expressed on the surface of activated platelets, PHA-activated lymphocytes and some tumor cell lines, including U937 and KG1a. LAMP-1 has been shown to be a ligand for E-selectin-mediated cell adhesion. LAMP-1 and LAMP-2 (CD107b) are carriers for poly-N-acetyllactosamines, and are able to display sialyl Le[x] termini.
Host Species: Mouse
Clone: H4A3
Isotype: IgG1, κ
Cross Reactivity: Human
Flow Cytometry
Specifications
CD107a | |
Flow Cytometry | |
APC | |
RUO | |
Human Adult Adherent Peripheral Blood Cells | |
Primary |
H4A3 | |
Monoclonal | |
Mouse | |
Human | |
IgG1 κ | |
Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.