Learn More
STEMCELL Technologies Doxorubicin (Hydrochloride), Size: 5 mg
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: STEMCELL Technologies 1000558
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Doxorubicin is an anthracycline antibiotic that intercalates DNA and inhibits DNA topoisomerase II (Burridge et al.; Lorenzo et al.). Doxorubicin exhibits cardiotoxic effects that involve generating reactive oxygen species, inhibiting DNA topoisomerase II, and releasing calcium in the sarcoplasmic reticulum (Burridge et al.). This product is supplied as the hydrochloride salt of the molecule.CANCER RESEARCH· Induces apoptosis in human endothelial cells (Lorenzo et al.).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.