Learn More
Ansul™ SPILL-X™ Agents in Shaker Bottles
Free-flowing dry agent to treat spills
$215.90 - $643.25
Alert:
The amount of spilled material neutralized or solidified depends on type of acid, caustic, solvent, or formaldehyde and its percentage concentration. Treated spills containing heavy metals or other listed wastes may still be characterized as hazardous. Disposal of spill waste materials should follow local, state and federal regulations.
Catalog Number | Mfr. No. | dimType | ForUseWithApplication | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | dimType | ForUseWithApplication | Price | Quantity & Availability | ||||
17-200-2A
|
Ansul™
077255 |
Pack of 6 for $215.90
|
|
||||||
17-200-2B
|
Ansul™
077265 |
Pack of 6 for $643.25
|
|
||||||
17-200-2C
|
Ansul™
077261 |
Pack of 6 for $364.64
|
|
||||||
17-200-3
|
Ansul™
077358 |
Pack of 6 for $314.13
|
|
||||||
17-200-6
|
Ansul™
078435 |
Pack of 6 for $351.87
|
|
||||||
Description
Free-flowing dry agent to treat spills
- Four agents available: SPILL-X-A™ for acids, SPILL-X-C™ for caustics, SPILL-X-S™ for solvents and SPILL-X-FP™ for formaldehyde
- Color-coded labels for easy identification
- Solidify and reduce vapor release from solvent or formaldehyde spills
- Neutralize and solidify acid or caustic spills
- Properly neutralized corrosive spills (pH over 2 and less than 12.5) are no longer characterized as “hazardous materials” by the U.S. EPA Properly treated solvent spills have greatly reduced vaporization loss
- Multi-agent kit includes two bottles each of SPILL-X-A, SPILL-X-C and SPILL-X-S agents
Order Info
Six single-agent shaker bottles per pack.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.