Learn More
Fine Science Tools ANGLED TIP REPLACEMENT
SDP
Supplier:  Fine Science Tools 1800002
Encompass
Product: Angled Tip for Small Vessel Cauterizer/ Tip Shape : Angled/ Tip Diameter : 0.3mm/ Alloy : Platinum-Iridium Unit : Each
Catalog No. NC9500933