Learn More
3B Scientific™ Angiospermae Fruits and Seeds Slide Set
15 microscope slides
Supplier: 3B Scientific™ W13051
Description
- 1(d). Triticum, wheat, t.s. of kernel (grain), endosperm and starch grains
- 2(e). Triticum, wheat, l.s. of kernel showing origin of embryo
- 3(d). Zea mays, corn, young cob t.s.
- 4(d). Phaseolus, bean, t.s. of pod with seed
- 5(d). Solanum, potato, t.s. of ovary with developing embryos
- 6(d). Helleborus, l.s. of atrope ovary
- 7(d). Capsella, l.s. of ovary with developing embryos
- 8(d). Papaver, poppy, t.s. of ovary with developing embryos
- 9(d). Phoenix, date-palm, t.s. of seed
- 10(d). Prunus, plum, t.s. of young stony fruit
- 11(d). Juglans regia, walnut, young drupe t.s.
- 12(d). Ribes, gooseberry, l.s. of young fruit
- 13(d). Helianthus, sunflower, t.s. of achene fruit
- 14(d). Pyrus malus, apple, young pome t.s., a fleshy, many seeded fruit
- 15(d). Fragaria, strawberry, young aggregate fruit l.s.
Specifications
Fruit/Seeds | |
15 |